Learn More
Waters Corp Column, Diol (OH), 5 μm, 3 mm × 250 mm
Catalog No. 502391591 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Torus column chemistries, including Torus Diol, allow for scaling SuperCritical Fluid Chromatography (SFC) from analytical to preparative applications. These columns offer a broad range of selectivity and are available in 1.7 μm and 5 μm analytical formats, as well as 5 μm preparative column formats.
  • Enables scaling SFC from analytical to preparative.
  • Column chemistries provide a wide range of selectivity.
  • Available in 1.7 μm and 5 μm analytical column formats.
  • Available in 5 μm preparative column formats.
Catalog No. 50-239-1591 Supplier Waters Corp Supplier No. 186008558
Torus column chemistries, including Torus Diol, allow for scaling SuperCritical Fluid Chromatography (SFC) from analytical to preparative applications. These columns offer a broad range of selectivity and are available in 1.7 μm and 5 μm analytical formats, as well as 5 μm preparative column formats.
  • Enables scaling SFC from analytical to preparative.
  • Column chemistries provide a wide range of selectivity.
  • Available in 1.7 μm and 5 μm analytical column formats.
  • Available in 5 μm preparative column formats.