Learn More
Waters Corp Column, Diol (OH), 1.7 μm, 3 mm x 75 mm
Catalog No. 502391433 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Torus Columns are designed to leverage the full capabilities of the ACQUITY UPC2 System for rapid, robust achiral separations. These columns streamline method development with four new 1.7 μm chemistries specifically for convergence chromatography. They deliver excellent peak shape, which minimizes or eliminates the need for additives, and provide enhanced selectivity for a wide array of compounds, along with improved robustness.
  • Designed for use with ACQUITY UPC2 System.
  • Facilitates fast, robust achiral separations.
  • Simplifies method development.
  • Features four innovative 1.7 μm chemistries for convergence chromatography.
  • Offers excellent peak shape, reducing or eliminating the need for additives.
  • Provides added selectivity for a wide range of compounds.
  • Ensures improved robustness.
Catalog No. 50-239-1433 Supplier Waters Corp Supplier No. 186007610
Torus Columns are designed to leverage the full capabilities of the ACQUITY UPC2 System for rapid, robust achiral separations. These columns streamline method development with four new 1.7 μm chemistries specifically for convergence chromatography. They deliver excellent peak shape, which minimizes or eliminates the need for additives, and provide enhanced selectivity for a wide array of compounds, along with improved robustness.
  • Designed for use with ACQUITY UPC2 System.
  • Facilitates fast, robust achiral separations.
  • Simplifies method development.
  • Features four innovative 1.7 μm chemistries for convergence chromatography.
  • Offers excellent peak shape, reducing or eliminating the need for additives.
  • Provides added selectivity for a wide range of compounds.
  • Ensures improved robustness.