Learn More
Waters Corp Torus Diol (OH) Column, 130Å, 1.7μm, 2.1 x 150 mm
Catalog No. 502392551 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Torus columns are designed for the ACQUITY UPC2 System, enabling fast, robust achiral separations. They simplify method development with four innovative 1.7 μm chemistries for convergence chromatography, providing excellent peak shape, reducing additive needs, and offering enhanced selectivity and improved robustness.
  • Specifically engineered to utilize the full range of capabilities of the ACQUITY UPC2 System.
  • Enables fast and robust achiral separations.
  • Simplifies the method development process.
  • Features four completely new and innovative 1.7 μm chemistries for convergence chromatography.
  • Designed for excellent peak shape.
  • Eliminates or reduces the need for additives.
  • Offers added selectivity for a wide range of compounds.
  • Provides improved robustness.
Catalog No. 50-239-2551 Supplier Waters Corp Supplier No. 186007608
Torus columns are designed for the ACQUITY UPC2 System, enabling fast, robust achiral separations. They simplify method development with four innovative 1.7 μm chemistries for convergence chromatography, providing excellent peak shape, reducing additive needs, and offering enhanced selectivity and improved robustness.
  • Specifically engineered to utilize the full range of capabilities of the ACQUITY UPC2 System.
  • Enables fast and robust achiral separations.
  • Simplifies the method development process.
  • Features four completely new and innovative 1.7 μm chemistries for convergence chromatography.
  • Designed for excellent peak shape.
  • Eliminates or reduces the need for additives.
  • Offers added selectivity for a wide range of compounds.
  • Provides improved robustness.