Learn More
Waters Corp Torus DIOL Column, 5 μm, 4.6 mm × 100 mm
Catalog No. 502392558 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Torus DIOL Columns were developed to provide additional selectivity choices within the Torus Column family. The high density Diol surface bonding offers chromatography similar to that from the traditional unbonded silica phases, but adds overall method robustness when utilized with additives.
  • Offers chromatography similar to that from traditional unbonded silica phases
  • Adds overall method robustness when utilized with additives
  • Shows good peak shapes for acidic compounds
  • Demonstrated by the separation of 6 isomeric forms of dimethoxybenzoic acid
Catalog No. 50-239-2558 Supplier Waters Corp Supplier No. 186008560
Torus DIOL Columns were developed to provide additional selectivity choices within the Torus Column family. The high density Diol surface bonding offers chromatography similar to that from the traditional unbonded silica phases, but adds overall method robustness when utilized with additives.
  • Offers chromatography similar to that from traditional unbonded silica phases
  • Adds overall method robustness when utilized with additives
  • Shows good peak shapes for acidic compounds
  • Demonstrated by the separation of 6 isomeric forms of dimethoxybenzoic acid