Learn More
Waters Corp Column, Diol (OH), 1.7 μm, 2.1 mm X 50 mm
Catalog No. 502391094 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Torus Columns are designed for fast, robust achiral separations on the ACQUITY UPC2 System. They simplify method development with innovative 1.7 μm chemistries, delivering excellent peak shapes and enhanced selectivity for a wide range of compounds.
  • Fast, robust achiral separations with ACQUITY UPC2 System.
  • Simplify method development using innovative 1.7 μm chemistries.
  • Excellent peak shapes and reduced need for additives.
  • Enhanced selectivity and robustness for diverse compounds.
  • High-density Diol (OH) surface bonding increases selectivity and method resilience.
  • Manufactured in an ISO-certified facility with ultra-pure reagents.
  • Each batch and column individually tested for repeatable performance.
  • Adjustable selectivity platform with two-stage functionalization.
  • Use of a Diol (OH) VanGuard pre-column recommended for protection.
Catalog No. 50-239-1094 Supplier Waters Corp Supplier No. 186007605
Torus Columns are designed for fast, robust achiral separations on the ACQUITY UPC2 System. They simplify method development with innovative 1.7 μm chemistries, delivering excellent peak shapes and enhanced selectivity for a wide range of compounds.
  • Fast, robust achiral separations with ACQUITY UPC2 System.
  • Simplify method development using innovative 1.7 μm chemistries.
  • Excellent peak shapes and reduced need for additives.
  • Enhanced selectivity and robustness for diverse compounds.
  • High-density Diol (OH) surface bonding increases selectivity and method resilience.
  • Manufactured in an ISO-certified facility with ultra-pure reagents.
  • Each batch and column individually tested for repeatable performance.
  • Adjustable selectivity platform with two-stage functionalization.
  • Use of a Diol (OH) VanGuard pre-column recommended for protection.