Learn More
Waters Corp Waters Torus Column, Diol (OH), 130Å, 5 μm, 4.6 mm x 50 mm
Catalog No. 502038648 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These columns are designed to streamline Supercritical Fluid Chromatography (SFC) from analytical to preparative scales. They offer a wide range of selectivity and are available in multiple particle sizes and column formats, including 1.7 μm and 5 μm analytical, and 5 μm preparative options.
  • Streamlines Supercritical Fluid Chromatography (SFC) from analytical to preparative scales
  • Offers a wide range of selectivity
  • Diol (OH) chemistry for SFC separation mode
  • Hybrid particle substrate with spherical shape
  • Temperature limits up to 60°C and maximum pressure of 6000 psi (415 Bar)
  • 5 μm particle size and 130 Å pore size
  • 4.6 mm inner diameter and 50 mm length
Catalog No. 50-203-8648 Supplier Waters Corp Supplier No. 186008559
These columns are designed to streamline Supercritical Fluid Chromatography (SFC) from analytical to preparative scales. They offer a wide range of selectivity and are available in multiple particle sizes and column formats, including 1.7 μm and 5 μm analytical, and 5 μm preparative options.
  • Streamlines Supercritical Fluid Chromatography (SFC) from analytical to preparative scales
  • Offers a wide range of selectivity
  • Diol (OH) chemistry for SFC separation mode
  • Hybrid particle substrate with spherical shape
  • Temperature limits up to 60°C and maximum pressure of 6000 psi (415 Bar)
  • 5 μm particle size and 130 Å pore size
  • 4.6 mm inner diameter and 50 mm length