Learn More
Waters Corp Torus OBD Prep Column, Diol (OH), 5 μm, 130 Å, 50 mm × 100 mm
Catalog No. 502392322 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters Torus Diol (OH) OBD Prep Columns are designed to scale SuperCritical Fluid (SFC) applications from analytical to preparative. Torus column chemistries offer a wide range of selectivity and are available in various analytical and preparative formats.
  • Scale supercritical fluid (SFC) from analytical to preparative.
  • Column chemistries offer a wide range of selectivity.
  • Available in 1.7 μm and 5 μm analytical and 5 μm preparative column formats.
Catalog No. 50-239-2322 Supplier Waters Corp Supplier No. 186008612
Add to Cart
Add to Cart
Request a Quote
Waters Torus Diol (OH) OBD Prep Columns are designed to scale SuperCritical Fluid (SFC) applications from analytical to preparative. Torus column chemistries offer a wide range of selectivity and are available in various analytical and preparative formats.
  • Scale supercritical fluid (SFC) from analytical to preparative.
  • Column chemistries offer a wide range of selectivity.
  • Available in 1.7 μm and 5 μm analytical and 5 μm preparative column formats.