Learn More
Waters Corp Torus Diol (OH) OBD Prep Column, 130Å, 5 μm, 50 mm × 150 mm
Catalog No. 502392797 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Torus column chemistries, including Diol, are designed to scale SuperCritical Fluid (SFC) from analytical to preparative. These chemistries offer a wide range of selectivity and are available in 1.7 μm and 5 μm analytical and 5 μm preparative column formats.
  • Scale SFC from analytical to preparative.
  • Offers a wide range of selectivity.
Catalog No. 50-239-2797 Supplier Waters Corp Supplier No. 186008613
Add to Cart
Add to Cart
Request a Quote
Torus column chemistries, including Diol, are designed to scale SuperCritical Fluid (SFC) from analytical to preparative. These chemistries offer a wide range of selectivity and are available in 1.7 μm and 5 μm analytical and 5 μm preparative column formats.
  • Scale SFC from analytical to preparative.
  • Offers a wide range of selectivity.