Learn More
Waters Corp Waters OBD Prep Column, Diol (OH), 5 μm, 19 mm x 50 mm
Catalog No. 502392444 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Torus DIOL Columns provide additional selectivity choices. The high-density diol surface bonding offers chromatography performance similar to traditional, unbonded silica phases, and enhances overall method robustness when utilized with additives.
  • Excellent peak shapes
  • Wide range of unique selectivities with unique ligands
  • Highest efficiency and QC-ready robustness
  • OBD Technology
  • Designed for achiral SFC separations
  • Ideally suited for method transfer and method scale-up
  • Based on patented two-stage functionalization of ethylene bridged hybrid (BEH) particles
  • Initial bonding provides a hydrophilic surface that controls retention characteristics and minimizes unwanted surface interactions
  • Robust chromatographic performance over the lifetime of the column
  • Show good peak shapes for acidic compounds
Catalog No. 50-239-2444 Supplier Waters Corp Supplier No. 186008602
Torus DIOL Columns provide additional selectivity choices. The high-density diol surface bonding offers chromatography performance similar to traditional, unbonded silica phases, and enhances overall method robustness when utilized with additives.
  • Excellent peak shapes
  • Wide range of unique selectivities with unique ligands
  • Highest efficiency and QC-ready robustness
  • OBD Technology
  • Designed for achiral SFC separations
  • Ideally suited for method transfer and method scale-up
  • Based on patented two-stage functionalization of ethylene bridged hybrid (BEH) particles
  • Initial bonding provides a hydrophilic surface that controls retention characteristics and minimizes unwanted surface interactions
  • Robust chromatographic performance over the lifetime of the column
  • Show good peak shapes for acidic compounds