Learn More
Waters Corp Torus DIOL Preparative Column, 130 Å, 5 μm, 30 x 75 mm
Catalog No. 502390548 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Torus DIOL Columns are designed for achiral SFC separations, providing additional selectivity options. They feature high-density diol surface bonding and are built on OBD Technology, developed using a patent-pending two-stage functionalization of ethylene bridged hybrid (BEH) particles. This design ensures robust chromatographic performance and suitability for method transfer and scale-up applications.
  • Excellent peak shapes
  • Wide range of unique selectivities with unique ligands
  • Highest efficiency and QC-ready robustness
  • Utilizes OBD Technology
  • Designed for achiral SFC separations
  • High-density diol surface bonding for robust chromatography
  • Maintains robust chromatographic performance over the lifetime of the column
Catalog No. 50-239-0548 Supplier Waters Corp Supplier No. 186008607
Add to Cart
Add to Cart
Request a Quote
Torus DIOL Columns are designed for achiral SFC separations, providing additional selectivity options. They feature high-density diol surface bonding and are built on OBD Technology, developed using a patent-pending two-stage functionalization of ethylene bridged hybrid (BEH) particles. This design ensures robust chromatographic performance and suitability for method transfer and scale-up applications.
  • Excellent peak shapes
  • Wide range of unique selectivities with unique ligands
  • Highest efficiency and QC-ready robustness
  • Utilizes OBD Technology
  • Designed for achiral SFC separations
  • High-density diol surface bonding for robust chromatography
  • Maintains robust chromatographic performance over the lifetime of the column