Learn More
Waters Corp Preparative Achiral SFC Column, DIOL, 130Å, 5 μm, 50 x 50 mm
Catalog No. 502391015 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Torus DIOL Columns enhance achiral SFC separations by offering additional selectivity and robust chromatographic performance. They feature a high-density diol surface and are suitable for method transfer and scale-up.
  • Enhances achiral SFC separations with added selectivity.
  • High-density diol surface similar to unbonded silica.
  • Increases method robustness.
  • Ensures good peak shapes for acidic compounds.
  • Suitable for method transfer and scale-up.
  • Features two-stage functionalization of BEH particles.
  • Hydrophilic surface controls retention, minimizes unwanted interactions.
  • Second functionalization step provides selectivity and peak shape.
  • Delivers broad selectivities and robust performance.
Catalog No. 50-239-1015 Supplier Waters Corp Supplier No. 186008611
Add to Cart
Add to Cart
Request a Quote
Torus DIOL Columns enhance achiral SFC separations by offering additional selectivity and robust chromatographic performance. They feature a high-density diol surface and are suitable for method transfer and scale-up.
  • Enhances achiral SFC separations with added selectivity.
  • High-density diol surface similar to unbonded silica.
  • Increases method robustness.
  • Ensures good peak shapes for acidic compounds.
  • Suitable for method transfer and scale-up.
  • Features two-stage functionalization of BEH particles.
  • Hydrophilic surface controls retention, minimizes unwanted interactions.
  • Second functionalization step provides selectivity and peak shape.
  • Delivers broad selectivities and robust performance.