Learn More
Waters Corp OBD Prep Column, DIOL Chemistry, 5 µm, 10 mm x 100 mm
Catalog No. 502391088 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Torus DIOL OBD Prep Column is engineered for robust achiral SFC separations, offering diverse selectivity and method scalability. Its high-density DIOL surface bonding provides excellent chromatography, enhancing method robustness for a broad range of analytes. It supports seamless method transfer and scale-up, utilizing two-stage functionalization of BEH particles for optimized retention.
  • Additional selectivity choices
  • High density DIOL surface bonding for chromatography
  • Increases method robustness
  • Good peak shapes for acidic compounds
  • Designed for achiral SFC separations
  • Wide selectivity and excellent peak shape
  • Suited for method transfer and scale-up
  • Optimized retention characteristics
  • Enhanced selectivity and peak shape
Catalog No. 50-239-1088 Supplier Waters Corp Supplier No. 186008599
The Torus DIOL OBD Prep Column is engineered for robust achiral SFC separations, offering diverse selectivity and method scalability. Its high-density DIOL surface bonding provides excellent chromatography, enhancing method robustness for a broad range of analytes. It supports seamless method transfer and scale-up, utilizing two-stage functionalization of BEH particles for optimized retention.
  • Additional selectivity choices
  • High density DIOL surface bonding for chromatography
  • Increases method robustness
  • Good peak shapes for acidic compounds
  • Designed for achiral SFC separations
  • Wide selectivity and excellent peak shape
  • Suited for method transfer and scale-up
  • Optimized retention characteristics
  • Enhanced selectivity and peak shape