Learn More
Carbosynth LLC. Tos-Arg-OMe HCl [TAME], 5g, MW: 342.41 36.46, Molecular formula: C14H22N4O4S HCI, CAS# 1784-03-8
SDP
Supplier:  Carbosynth LLC. STR30035G
Encompass_Preferred
This item has a minimum order quantity of 4 per supplier requirements
Synonyms: [TAME] Tosyl-L-Arginine Methyl Ester Monohydrochloride, Application: Substrate for Trypsin, Storage Temperature: -20C, References: K.A. Walsh, In, Proteolytic Enzymes, Methods in Enzymology, Vol. 19, (G.E. Perlmann and L. Lorand, Eds.), Academic Press, New York, 1970, pp. 41-63.
Catalog No. 50-168-7169