Learn More
Bachem Tos-Gly-Pro-Arg-AMC · HCl
SDP
Catalog No. 501947044
Encompass_Preferred
Tos-Gly-Pro-Arg-AMC · HCl, 117961-27-0, C30H37N7O7S · HCl, Mr 676.19, 50mg. Tos-GPR-AMC, fluorogenic substrate for trypsin and thrombin. Kinetic data for Thrombin: kcat = 180, Km = 310 uM.
Catalog No. 50-194-7044 Supplier Bachem Supplier No. 4008607.0050
Add to Cart
Add to Cart
Tos-Gly-Pro-Arg-AMC · HCl, 117961-27-0, C30H37N7O7S · HCl, Mr 676.19, 50mg. Tos-GPR-AMC, fluorogenic substrate for trypsin and thrombin. Kinetic data for Thrombin: kcat = 180, Km = 310 uM.