Learn More
Bachem Tos-Lys-chloromethylketone · HCl
SDP
Supplier:  Bachem 4001453.0500
Encompass_Preferred
Tos-Lys-chloromethylketone · HCl, 4272-74-6, C14H21ClN2O3S · HCl, Mr 369.31, 500mg. TLCK. Tos-Lys-CMK, TLCK, a highly effective irreversible inhibitor of lysyl endopeptidase. TLCK and TPCK (N-1160) are potent inhibitors of activated caspase proteases. CAS Number (net): 2364-87-6.
Catalog No. 50-397-132