Learn More
Medchemexpress LLC Tos-PEG1-CH2-Boc | 1643957-24-7 | 100 MG
SDP
Catalog No. 5000339936
Encompass_Preferred
Tos-PEG1-CH2-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein, exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used in the synthesis of PROTACs
  • PEG-based PROTAC linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-39936 Supplier Medchemexpress LLC Supplier No. HY130158100MG
Tos-PEG1-CH2-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein, exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used in the synthesis of PROTACs
  • PEG-based PROTAC linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins