Learn More
Medchemexpress LLC Tos-PEG3 | 77544-68-4 | 97.0% | 304.36 | 1 G
SDP
Supplier:  Medchemexpress LLC HY234081G
Encompass_Preferred
Tos-PEG3 is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs. It can also be used for the synthesis of 3'-aminooxy oligonucleotides solid supports.
  • Can be used in the synthesis of PROTACs
  • Can be utilized for the synthesis of 3'-aminooxy oligonucleotides solid supports
Catalog No. 50-004-02579