Learn More
Medchemexpress LLC Tos-PEG3 | 77544-68-4 | 97.0% | 304.36 | 10 G
SDP
Catalog No. 5000400968
Encompass_Preferred
Tos-PEG3 is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs. It can also be used for the synthesis of 3'-aminooxy oligonucleotides solid supports.
  • Utilized in the synthesis of PROTACs.
  • Can be used for the synthesis of 3'-aminooxy oligonucleotides solid supports.
Catalog No. 50-004-00968 Supplier Medchemexpress LLC Supplier No. HY2340810G
Tos-PEG3 is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs. It can also be used for the synthesis of 3'-aminooxy oligonucleotides solid supports.
  • Utilized in the synthesis of PROTACs.
  • Can be used for the synthesis of 3'-aminooxy oligonucleotides solid supports.