Learn More
Medchemexpress LLC Tos-PEG3 | 77544-68-4 | 97.0% | 304.36 | 50 G
SDP
Catalog No. 5000400867
Encompass_Preferred
Tos-PEG3 is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It can also be utilized for the synthesis of 3'-aminooxy oligonucleotides solid supports.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Can be utilized for the synthesis of 3'-aminooxy oligonucleotides solid supports
Catalog No. 50-004-00867 Supplier Medchemexpress LLC Supplier No. HY2340850G
Tos-PEG3 is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It can also be utilized for the synthesis of 3'-aminooxy oligonucleotides solid supports.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Can be utilized for the synthesis of 3'-aminooxy oligonucleotides solid supports