Learn More
Medchemexpress LLC Tos-peg3-ch2cooh | 1581248-63-6 | 100 MG
SDP
Catalog No. 5000340534
Encompass_Preferred
Tos-PEG3-CH2COOH is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. PROTACs are molecules that leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins. They consist of two different ligands connected by a linker; one ligand binds to an E3 ubiquitin ligase, and the other binds to the target protein, facilitating its degradation.
  • Utilizes the ubiquitin-proteasome system for selective protein degradation.
  • Acts as a linker in the construction of degraders.
Catalog No. 50-003-40534 Supplier Medchemexpress LLC Supplier No. HY130674100MG
Tos-PEG3-CH2COOH is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. PROTACs are molecules that leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins. They consist of two different ligands connected by a linker; one ligand binds to an E3 ubiquitin ligase, and the other binds to the target protein, facilitating its degradation.
  • Utilizes the ubiquitin-proteasome system for selective protein degradation.
  • Acts as a linker in the construction of degraders.