Learn More
Medchemexpress LLC Tos-PEG3-CH2COOH | 1581248-63-6 | 50 MG
SDP
Catalog No. 5000339865
Encompass_Preferred
Tos-PEG3-CH2COOH is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These PROTACs feature two distinct ligands connected by a linker: one targets an E3 ubiquitin ligase, and the other binds to a specific target protein. This mechanism allows PROTACs to exploit the intracellular ubiquitin-proteasome system for the selective degradation of target proteins. This product is for research use only.
  • PEG-based PROTAC linker
  • Used for PROTAC synthesis
  • Leverages ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-39865 Supplier Medchemexpress LLC Supplier No. HY13067450MG
Tos-PEG3-CH2COOH is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These PROTACs feature two distinct ligands connected by a linker: one targets an E3 ubiquitin ligase, and the other binds to a specific target protein. This mechanism allows PROTACs to exploit the intracellular ubiquitin-proteasome system for the selective degradation of target proteins. This product is for research use only.
  • PEG-based PROTAC linker
  • Used for PROTAC synthesis
  • Leverages ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only