Learn More
Medchemexpress LLC Tos-PEG3-CH2COOH | 1581248-63-6 | 98.0% | 362.4 g·mol⁻¹ | C15H22O8S | 5 MG
SDP
Catalog No. 5000262156
Encompass_Preferred
Tos-PEG3-CH2COOH is a polyethylene glycol (PEG)-based linker bearing a tosylated PEG3 spacer and a terminal carboxylic acid group. It is intended for use in medicinal chemistry workflows as a spacer/conjugation handle during the synthesis of proteolysis-targeting chimeras (PROTACs). Consult the manufacturer's certificate of analysis for batch-specific handling and storage instructions.
  • Tosylated PEG3 spacer terminating in a carboxylic acid
  • Designed for use as a PROTAC linker and conjugation handle
  • Molecular weight 362.4 g·mol⁻¹; formula C15H22O8S
  • Available in small research pack sizes suitable for synthesis screening
  • Reported purity approximately 98% (supplier listing)
  • Store under conditions specified in the certificate of analysis
Catalog No. 50-002-62156 Supplier Medchemexpress LLC Supplier No. HY1306745MG
Tos-PEG3-CH2COOH is a polyethylene glycol (PEG)-based linker bearing a tosylated PEG3 spacer and a terminal carboxylic acid group. It is intended for use in medicinal chemistry workflows as a spacer/conjugation handle during the synthesis of proteolysis-targeting chimeras (PROTACs). Consult the manufacturer's certificate of analysis for batch-specific handling and storage instructions.
  • Tosylated PEG3 spacer terminating in a carboxylic acid
  • Designed for use as a PROTAC linker and conjugation handle
  • Molecular weight 362.4 g·mol⁻¹; formula C15H22O8S
  • Available in small research pack sizes suitable for synthesis screening
  • Reported purity approximately 98% (supplier listing)
  • Store under conditions specified in the certificate of analysis