Learn More
Medchemexpress LLC Tos-PEG3-CH2COOH | 1581248-63-6 | C15H22O8S | 10MG
SDP
Catalog No. 5000259785
Encompass_Preferred
Tos-PEG3-CH2COOH is a PEG-based PROTAC linker with a tosyl cap and a three-unit polyethylene glycol spacer terminating in a carboxylic acid. It is designed for use in the synthesis of PROTACs and other bifunctional molecules where a short, flexible, polar spacer and a carboxyl functional handle are required.
  • Tosyl-capped PEG3 spacer ending in a carboxylic acid for conjugation.
  • Provides a short, flexible, hydrophilic linker suitable for cellular applications.
  • Terminal carboxyl group enables amide or ester coupling chemistries.
  • Compatible with solid-phase and solution-phase synthetic workflows.
  • Supplied in research-scale quantities for medicinal chemistry and PROTAC development.
  • Molecular formula C15H22O8S, molecular weight ≈362.40 g/mol.
Catalog No. 50-002-59785 Supplier Medchemexpress LLC Supplier No. HY13067410MG
Tos-PEG3-CH2COOH is a PEG-based PROTAC linker with a tosyl cap and a three-unit polyethylene glycol spacer terminating in a carboxylic acid. It is designed for use in the synthesis of PROTACs and other bifunctional molecules where a short, flexible, polar spacer and a carboxyl functional handle are required.
  • Tosyl-capped PEG3 spacer ending in a carboxylic acid for conjugation.
  • Provides a short, flexible, hydrophilic linker suitable for cellular applications.
  • Terminal carboxyl group enables amide or ester coupling chemistries.
  • Compatible with solid-phase and solution-phase synthetic workflows.
  • Supplied in research-scale quantities for medicinal chemistry and PROTAC development.
  • Molecular formula C15H22O8S, molecular weight ≈362.40 g/mol.