Learn More
Medchemexpress LLC Tos-PEG3-CH2COOtBu | 1530778-24-5 | 418.50 | 10 G
SDP
Catalog No. 5000402131
Encompass_Preferred
Tos-PEG3-CH2COOtBu is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Contains two different ligands connected by a linker
  • Ligands for an E3 ubiquitin ligase and a target protein
Catalog No. 50-004-02131 Supplier Medchemexpress LLC Supplier No. HY4501810G
Tos-PEG3-CH2COOtBu is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Contains two different ligands connected by a linker
  • Ligands for an E3 ubiquitin ligase and a target protein