Learn More
Medchemexpress LLC Tos-PEG3-NH-Boc (PROTAC Linker 9) | 206265-94-3 | 99.7% | 250 MG
SDP
Catalog No. 5000341530
Encompass_Preferred
Tos-PEG3-NH-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are compounds containing two different ligands connected by a linker: one for an E3 ubiquitin ligase and the other for a target protein. They utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.
  • PEG-based linker
  • Used in the synthesis of PROTACs
  • Supports PROTACs that selectively degrade target proteins
  • Liquid appearance
  • Colorless to light yellow
Catalog No. 50-003-41530 Supplier Medchemexpress LLC Supplier No. 250MGHY128805
Tos-PEG3-NH-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are compounds containing two different ligands connected by a linker: one for an E3 ubiquitin ligase and the other for a target protein. They utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.
  • PEG-based linker
  • Used in the synthesis of PROTACs
  • Supports PROTACs that selectively degrade target proteins
  • Liquid appearance
  • Colorless to light yellow