Learn More
Medchemexpress LLC Tos-PEG4-acid | 1453117-42-4 | 97.2% | 250 MG
SDP
Catalog No. 5000341454
Encompass_Preferred
Tos-PEG4-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Enables PROTACs to contain two different ligands connected by a linker
  • Facilitates targeting of E3 ubiquitin ligase and target proteins
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-41454 Supplier Medchemexpress LLC Supplier No. 250MGHY130417
Tos-PEG4-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Enables PROTACs to contain two different ligands connected by a linker
  • Facilitates targeting of E3 ubiquitin ligase and target proteins
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins