Learn More
Medchemexpress LLC Tos-PEG4-acid | 1453117-42-4 | 97.2% | 50 MG
SDP
Catalog No. 5000340560
Encompass_Preferred
Tos-PEG4-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. This compound facilitates the degradation of target proteins by exploiting the intracellular ubiquitin-proteasome system.
  • Peg-based PROTAC linker
  • Utilized in PROTAC synthesis
  • Facilitates protein degradation
  • Selectively targets proteins via ubiquitin-proteasome system
Catalog No. 50-003-40560 Supplier Medchemexpress LLC Supplier No. HY13041750MG
Tos-PEG4-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. This compound facilitates the degradation of target proteins by exploiting the intracellular ubiquitin-proteasome system.
  • Peg-based PROTAC linker
  • Utilized in PROTAC synthesis
  • Facilitates protein degradation
  • Selectively targets proteins via ubiquitin-proteasome system