Learn More
Medchemexpress LLC HY-42620 250mg Medchemexpress, Tos-PEG4-CH2-Boc CAS:169751-73-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY42620250MG
Encompass_Preferred
Medchemexpress, HY-42620 250mg Tos-PEG4-CH2-Boc CAS:169751-73-9 Tos-PEG4-CH2-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2242