Learn More
Medchemexpress LLC Tos-peg4-nh-boc (PROTAC linker 7) | 1246999-33-6 | ≥98.0% | 1 G
SDP
Catalog No. 5000340546
Encompass_Preferred
Tos-PEG4-NH-Boc (PROTAC Linker 7) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker; one ligand is for an E3 ubiquitin ligase, and the other is for the target protein. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Consists of two different ligands connected by a linker
  • Utilizes the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-40546 Supplier Medchemexpress LLC Supplier No. HY1288031G
Tos-PEG4-NH-Boc (PROTAC Linker 7) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker; one ligand is for an E3 ubiquitin ligase, and the other is for the target protein. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Consists of two different ligands connected by a linker
  • Utilizes the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins