Learn More
Medchemexpress LLC Tos-PEG4-NH-Boc (PROTAC Linker 7) | 1246999-33-6 | ≥98.0% | 250 MG
SDP
Catalog No. 5000341469
Encompass_Preferred
Tos-PEG4-NH-Boc is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These molecules are designed to leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Utilizes the ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Consists of two different ligands connected by a linker
  • One ligand binds to an E3 ubiquitin ligase
  • Other ligand binds to the target protein
Catalog No. 50-003-41469 Supplier Medchemexpress LLC Supplier No. 250MGHY128803
Tos-PEG4-NH-Boc is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These molecules are designed to leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Utilizes the ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Consists of two different ligands connected by a linker
  • One ligand binds to an E3 ubiquitin ligase
  • Other ligand binds to the target protein