Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tos-PEG4-t-butyl ester | 217817-01-1 | 50 MG
SDP

Catalog No. 5000340244
Encompass_Preferred

Tos-PEG4-t-butyl ester is a PROTAC linker with a PEG composition. It is utilized in the synthesis of various PROTACs.

  • Molecular weight: 432.53
  • Chemical formula: C20H32O8S
  • Appearance: Liquid
  • Color: Colorless to light yellow

Catalog No. 50-003-40244 Supplier Medchemexpress LLC Supplier No. HY13042250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tos-PEG4-t-butyl ester is a PROTAC linker with a PEG composition. It is utilized in the synthesis of various PROTACs.

  • Molecular weight: 432.53
  • Chemical formula: C20H32O8S
  • Appearance: Liquid
  • Color: Colorless to light yellow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.