Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tos-PEG4-THP | 86259-89-4 | 250 MG
SDP

Catalog No. 5000342470
Encompass_Preferred

Tos-PEG4-THP is a PEG-based PROTAC linker designed for use in the synthesis of PROTAC K-Ras Degrader-1. PROTACs are compounds that leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins, featuring two different ligands connected by a linker: one for an E3 ubiquitin ligase and another for the target protein.

  • Used in PROTAC synthesis
  • Targeted for PROTAC linkers and PROTAC pathway
  • Storage at -20°C, under nitrogen
  • Solvent storage at -80°C for 6 months; -20°C for 1 month (under nitrogen)
  • Soluble in H2O at 100 mg/mL

Catalog No. 50-003-42470 Supplier Medchemexpress LLC Supplier No. 250MGHY130819
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tos-PEG4-THP is a PEG-based PROTAC linker designed for use in the synthesis of PROTAC K-Ras Degrader-1. PROTACs are compounds that leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins, featuring two different ligands connected by a linker: one for an E3 ubiquitin ligase and another for the target protein.

  • Used in PROTAC synthesis
  • Targeted for PROTAC linkers and PROTAC pathway
  • Storage at -20°C, under nitrogen
  • Solvent storage at -80°C for 6 months; -20°C for 1 month (under nitrogen)
  • Soluble in H2O at 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.