Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tos-PEG5-Boc | 581065-94-3 | ≥97.0% | 1 G
SDP

Catalog No. 5000340835
Encompass_Preferred

Tos-PEG5-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. These compounds consist of two distinct ligands connected by a linker: one targeting an E3 ubiquitin ligase and the other targeting the protein intended for degradation. PROTACs leverage the intracellular ubiquitin-proteasome system for selective protein degradation.

  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Molecular weight: 476.58
  • Chemical formula: C22H36O9S
  • Appears as a viscous liquid
  • Colorless to light yellow

Catalog No. 50-003-40835 Supplier Medchemexpress LLC Supplier No. HY1305231G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tos-PEG5-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. These compounds consist of two distinct ligands connected by a linker: one targeting an E3 ubiquitin ligase and the other targeting the protein intended for degradation. PROTACs leverage the intracellular ubiquitin-proteasome system for selective protein degradation.

  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Molecular weight: 476.58
  • Chemical formula: C22H36O9S
  • Appears as a viscous liquid
  • Colorless to light yellow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.