Learn More
Medchemexpress LLC Tos-PEG5-Boc | 581065-94-3 | ≥97.0% | 250 MG
SDP
Catalog No. 5000341760
Encompass_Preferred
Tos-PEG5-Boc is a PEG-based PROTAC linker designed for the synthesis of PROTACs. These molecules exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting a ligand for an E3 ubiquitin ligase and a ligand for the target protein. It has a molecular weight of 476.58 and a chemical formula of C22H36O9S. Its appearance is a viscous liquid, colorless to light yellow.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Viscous liquid, colorless to light yellow appearance
  • For research use only
Catalog No. 50-003-41760 Supplier Medchemexpress LLC Supplier No. 250MGHY130523
Tos-PEG5-Boc is a PEG-based PROTAC linker designed for the synthesis of PROTACs. These molecules exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting a ligand for an E3 ubiquitin ligase and a ligand for the target protein. It has a molecular weight of 476.58 and a chemical formula of C22H36O9S. Its appearance is a viscous liquid, colorless to light yellow.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Viscous liquid, colorless to light yellow appearance
  • For research use only