Learn More
Medchemexpress LLC Tos-PEG5-Boc | 581065-94-3 | ≥97.0% | 5 G
SDP
Catalog No. 5000339968
Encompass_Preferred
Tos-PEG5-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting an E3 ubiquitin ligase ligand and a target protein ligand via a linker. It has a molecular weight of 476.58 and the formula C22H36O9S, appearing as a viscous liquid that is colorless to light yellow.
  • PEG-based PROTAC linker
  • Used for the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Viscous liquid, colorless to light yellow appearance
Catalog No. 50-003-39968 Supplier Medchemexpress LLC Supplier No. HY1305235G
Tos-PEG5-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting an E3 ubiquitin ligase ligand and a target protein ligand via a linker. It has a molecular weight of 476.58 and the formula C22H36O9S, appearing as a viscous liquid that is colorless to light yellow.
  • PEG-based PROTAC linker
  • Used for the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Viscous liquid, colorless to light yellow appearance