Learn More
Medchemexpress LLC Tos-PEG5-CH2COOtBu | 1949793-62-7 | C23H38O10S | 250 MG
SDP
Catalog No. 5000352549
Encompass_Preferred
Tos-PEG5-CH2COOtBu is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is associated with applications in cancer-programmed cell death, and its target is PROTAC Linkers.
  • Used in the synthesis of PROTACs.
  • Associated with applications in cancer-programmed cell death.
  • Target is PROTAC linkers.
Catalog No. 50-003-52549 Supplier Medchemexpress LLC Supplier No. 250MGHY140366
Tos-PEG5-CH2COOtBu is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is associated with applications in cancer-programmed cell death, and its target is PROTAC Linkers.
  • Used in the synthesis of PROTACs.
  • Associated with applications in cancer-programmed cell death.
  • Target is PROTAC linkers.