Learn More
Medchemexpress LLC Tos-PEG9 | 62573-11-9 | C25H44O12S | 250 MG
SDP
Catalog No. 5000352825
Encompass_Preferred
Tos-PEG9 is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs themselves consist of two different ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets the protein of interest. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-52825 Supplier Medchemexpress LLC Supplier No. 250MGHY140357
Tos-PEG9 is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs themselves consist of two different ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets the protein of interest. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins