Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TOS Titration Medium, Ricca Chemical
SDP

Catalog No. RR86605004C Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RR86605004C 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RR86605004C Supplier Ricca Chemical Company Supplier No. R86605004C
Add to Cart
Add to Cart

Specifications

Chemical Name or Material TOS titration medium
CAS 7647-01-0, 7732-18-5
Grade Laboratory
Packaging Amber Glass Bottle
pH Acidic
Quantity 4 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.