Learn More
Apexbio Technology LLC Tosedostat (CHR2797), 10mg. Cas: 238750-77-1 MFCD: MFCD13185162. Aminopeptidase inhibitor
SDP
Supplier:  Apexbio Technology LLC A435510
Encompass_Preferred
Tosedostat is a novel and potent oral aminopeptidase inhibitor with clinical activity in a previous phase 1-2 study in elderly patients with relapsed or refractory acute myeloid leukaemia (AML). [2]Aminopeptidases play a key role in the protein cell cycle. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4928