Learn More
Healthmark Industries Co TOSI PROFORMANCE WASHR TST 30C
SDP
Catalog No. NC0704006
Encompass_Preferred
TOSI® is the first device to provide a consistent, repeatable, and reliable method for evaluating the cleaning effectiveness of the automated instrument washer The routine use of this test will help insure that your instrument washer is performing at a consistent level, enhancing the routine visual inspection of instruments TOSI® complies with new AAMI and AORN Guidelines as well as ASTM Guide D7225 This product is required to be reported under California Proposition 65
Catalog No. NC0704006 Supplier Healthmark Industries Co Supplier No. WT101
TOSI® is the first device to provide a consistent, repeatable, and reliable method for evaluating the cleaning effectiveness of the automated instrument washer The routine use of this test will help insure that your instrument washer is performing at a consistent level, enhancing the routine visual inspection of instruments TOSI® complies with new AAMI and AORN Guidelines as well as ASTM Guide D7225 This product is required to be reported under California Proposition 65