Learn More
Enzo Life Sciences Tosufloxacin . tosylate (250mg). CAS: 115964-29-9
SDP
Supplier:  Enzo Life Sciences ALX380303M250
Encompass_Preferred
Fluoroquinolone antibacterial agent. Inhibits bacterial DNA gyrase (topoisomerase). Effective against Gram-positive and Gram-negative aerobic bacteria, anaerobic bacteria and Chlamydia trachomatis. Alternative name: 7-(3-Amino-1-pyrrolidinyl)-1-(2,4-difluorophenyl)-6-fluoro-1,4-dihydro-4-oxo-1,8-naphthyridine-3-carboxylic acid . tosylate, A-64730. Purity: ≥98% (HPLC). Solubility: Slightly soluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-9026