Learn More
Bioworld TOSYL-ACTIVATED SEPAROPORE
Catalog No. 50197763
Encompass
This item has a minimum order quantity of 2 per supplier requirements. TOSYL-ACTIVATED SEPAROPORE (AGAROSE) 4B-CL: Tosyl-activated Separopore 4B-CL Can be used to couple primary amines, thiols, imidazole or tyrosine hydroxyl groups. Tosyl-activated Separopore couples more rapidly and efficiently to sulfhyd...
Catalog No. 50-197-763 Supplier Bioworld Supplier No. 201810391
This item has a minimum order quantity of 2 per supplier requirements. TOSYL-ACTIVATED SEPAROPORE (AGAROSE) 4B-CL: Tosyl-activated Separopore 4B-CL Can be used to couple primary amines, thiols, imidazole or tyrosine hydroxyl groups. Tosyl-activated Separopore couples more rapidly and efficiently to sulfhyd...