Learn More
Bioworld TOSYL-ACTIVATED SEPAROPORE
Supplier:  Bioworld 201810392
Encompass
TOSYL-ACTIVATED SEPAROPORE (AGAROSE) 4B-CL: Tosyl-activated Separopore 4B-CL Can be used to couple primary amines, thiols, imidazole or tyrosine hydroxyl groups. Tosyl-activated Separopore couples more rapidly and efficiently to sulfhyd...
Catalog No. 50-197-764