Learn More
Selleck Chemical LLC Tosylic acid
SDP
Catalog No. 5000002978
Encompass_Preferred
p-Toluenesulfonic acid (4-Methylbenzenesulfonic acid Tosylic acid tosic acid para-toluene) is an extremely strong acidic compound
Catalog No. 50-000-02978 Supplier Selleck Chemical LLC Supplier No. S618825MG
p-Toluenesulfonic acid (4-Methylbenzenesulfonic acid Tosylic acid tosic acid para-toluene) is an extremely strong acidic compound