Learn More
Apexbio Technology LLC Tosyllysine Chloromethyl Ketone (hydrochloride) 4272-74-6 250mg
SDP
Catalog No. 5000089682
Encompass_Preferred
Tosyllysine Chloromethyl Ketone hydrochloride is a small-molecule inhibitor of serine proteases with trypsin-like activity acting through irreversible covalent modification of protease active sites It can also inhibit certain cysteine proteases and thiol-dependent enzymes via nonspecific reactivity with free thiols In cell-based assays it suppresses inducible nitric oxide synthase (iNOS) expression in stimulated macrophages and modulates protease-mediated cytotoxicity induced by bacterial factors These properties highlight its research value in protein extraction and purification regulation of cellular proteolysis and studies of protease-dependent cellular responses
Catalog No. 50-000-89682 Supplier Apexbio Technology LLC Supplier No. C5839250
Tosyllysine Chloromethyl Ketone hydrochloride is a small-molecule inhibitor of serine proteases with trypsin-like activity acting through irreversible covalent modification of protease active sites It can also inhibit certain cysteine proteases and thiol-dependent enzymes via nonspecific reactivity with free thiols In cell-based assays it suppresses inducible nitric oxide synthase (iNOS) expression in stimulated macrophages and modulates protease-mediated cytotoxicity induced by bacterial factors These properties highlight its research value in protein extraction and purification regulation of cellular proteolysis and studies of protease-dependent cellular responses