Learn More
Cayman Chemical Tosyllysine Chloromethyl Ketone (hydrochloride), Purity: ≥95%, CAS: 4272-74-6, 100 mg
SDP
Catalog No. 502955873
Encompass_Preferred
An active site-directed agent that inhibits serine proteinases with trypsin-like activity; may also act non-selectively with thiol groups and thereby inhibit cysteine proteinases and other enzymes
Catalog No. 50-295-5873 Supplier Cayman Chemical Supplier No. 13074100 MG
An active site-directed agent that inhibits serine proteinases with trypsin-like activity; may also act non-selectively with thiol groups and thereby inhibit cysteine proteinases and other enzymes