Learn More
Hach Company Total Inorganic Nitrogen Base Concentrate, 50 mL
Supplier:  Hach Company 204059
Encompass
For total inorganic nitrogen determination by the Titanium Trichloride Reduction method
Catalog No. 50-199-4665