Learn More
Norgen Biotek Corp Total RNA Purification 96-Well Kit
Catalog No. NC1809201
Encompass_Preferred
Extract high quality & purity total RNA including miRNA . Convenient high throughput format for vacuum, centrifuge or robot. Very consistent well-to-well RNA isolation. No phenol step required - isolate all RNA in one fraction. Bind & elute all RNA irrespective of size or GC content, without bias. Isolate from a wide variety of specimens.
Catalog No. NC1809201 Supplier Norgen Biotek Corp Supplier No. 24380
Extract high quality & purity total RNA including miRNA . Convenient high throughput format for vacuum, centrifuge or robot. Very consistent well-to-well RNA isolation. No phenol step required - isolate all RNA in one fraction. Bind & elute all RNA irrespective of size or GC content, without bias. Isolate from a wide variety of specimens.