Learn More
Crystal Chem Inc Total 25-OH Vitamin D ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80987
Encompass
The new, and updated, Vitamin D (25-OH) Total ELISA is a highly sensitive assay that is used to quantify levels of total vitamin D (25-OH) in plasma and serum. Highlights: No separate pretreatment. For use with plasma and serum. 100% Cross reactivity with 25-OH Vitamin D2/D3. Under 2hour assay time.
Catalog No. 50-194-7888