Learn More
Meso Scale Discovery Total Akt Kit
SDP
Catalog No. 5000080572
Encompass
The Meso Scale Discovery Total Akt Kit is a 96-well plate assay for measuring total Akt protein in cell and tissue lysates It uses MSD s SULFO-TAG labels and multi-array technology for a fast sensitive and reproducible quantification method The kit includes a streptavidin plate with a capture antibody a detection antibody with ruthenium sample diluent wash buffer read buffer and cell lysate controls It has a sensitivity of 1-2 ng/mL and a range up to 10 000 pg/mL compatible with various buffers and detergents ideal for high-throughput Akt screening
Catalog No. 50-000-80572 Supplier Meso Scale Discovery Supplier No. K150MOD1
The Meso Scale Discovery Total Akt Kit is a 96-well plate assay for measuring total Akt protein in cell and tissue lysates It uses MSD s SULFO-TAG labels and multi-array technology for a fast sensitive and reproducible quantification method The kit includes a streptavidin plate with a capture antibody a detection antibody with ruthenium sample diluent wash buffer read buffer and cell lysate controls It has a sensitivity of 1-2 ng/mL and a range up to 10 000 pg/mL compatible with various buffers and detergents ideal for high-throughput Akt screening